Rhodium, foil, Thickness: 0,1 mm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-16-6
:
Formula:Rh
MDL Number:MFCD00011201
Merck Index:12,08353
Boiling Pt:3400…3700 °C (1013 hPa)
Melting Pt:1960 °C
Density:12.41 g/cm³ (20 °C)
:102.9 g/mol
EINECS:231-125-0
Frequently Bought Together