Platinum, black, high surface area
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 7440-06-4 Physical Form: S.A. 25.0-30.4m2/g Odour: Odourless MDL No.: MFCD00011179
Synonyms: Platinium
- CAS Number:7440-06-4
- Formula:Pt
- MDL Number:MFCD00011179
- Molecular Weight:195.08 g/mol
- Boiling Pt:4530 °C (1013 hPa)
- Melting Pt:1773 °C
- Density:21.4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-116-1
- UN:3089
- ADR:4.1,II
2 Options Available Below
Product Details & Documents
CAS No.: 7440-06-4 Physical Form: S.A. 25.0-30.4m2/g Odour: Odourless MDL No.: MFCD00011179
Synonyms: Platinium
- CAS Number:7440-06-4
- Formula:Pt
- MDL Number:MFCD00011179
- Molecular Weight:195.08 g/mol
- Boiling Pt:4530 °C (1013 hPa)
- Melting Pt:1773 °C
- Density:21.4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-116-1
- UN:3089
- ADR:4.1,II
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice



