Palladium ≥99.9% (trace metals basis), sponge, Sigma-Aldrich®
SIAL267082-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:106.420 g/mol
:12,07121
:3167 °C (1013 hPa)
:1555 °C
:12.02 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:231-115-6
:7440-05-3
:Pd
:MFCD00011167