Poly(caprolactone), M.W. 80000, pellets ~3 mm, Sigma-Aldrich®
: Merck
:24980-41-4
:
:(C₆H₁₀O₂)n
:MFCD00084404
:114.1400 g/mol
:60 °C (lit.)
:1.145 g/ml (25 °C)
:M-Clarity™ quality level = MQ200
- Infrared spectrum:Conforms to structure
- Water content (impurities):<0.5%
- Melt flow index (160 degrees celsius/5.00 kg):2.01 - 4.03
- Molecular weight:Average Mn 80000
Frequently Bought Together