Tantalum, powder, Sigma-Aldrich®
Catalog # SIAL545007-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:180.95 g/mol
Density:16.6 g/cm³ (20 °C)
UN:3089
ADR:4.1,II
:M-Clarity™ quality level = MQ100
EINECS:231-135-5
:GHS02
CAS Number:7440-25-7
Formula:Ta
MDL Number:MFCD00011252