Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-20-2
:
Formula:Sc
MDL Number:MFCD00016323
:44.96 g/mol
Merck Index:12,08537
Boiling Pt:2730 °C (1013 hPa)
Melting Pt:1541 °C
Density:2.989 g/cm³ (20 °C)
:Packaged under argon.
Storage Temperature:Ambient
EINECS:231-129-2
Frequently Bought Together