Zinc, powder, Sigma-Aldrich®
Catalog # SIAL578002-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:65.39 g/mol
Boiling Pt:907 °C (1013 hPa)
Melting Pt:419.5 °C
Density:7.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
UN:1436
ADR:4.3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS09
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291