Order Entry
United Kingdom
ContactUsLinkComponent
Mercury
Toxic
Health
Environment
Mercury
Catalog # SIALERMAE640-5ML
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
Mercury
Catalog # SIALERMAE640-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :200.59 g/mol
Merck Index:13,05925
Boiling Pt:356.5 °C (1013 hPa)
Melting Pt:–39 °C
Density:13.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-106-7
UN:2809
ADR:8,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:7439-97-6
Formula:Hg
MDL Number:MFCD00011035