Order Entry
United Kingdom
ContactUsLinkComponent
Cadmium ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Cadmium ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Catalog # 44127.FI
CAS Number:  
Cadmium ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Catalog # 44127.FI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1670
  • Pk Info:
    50×50 mm

Specifications

About this item

CAS Number:7440-43-9
 :
Formula:Cd
MDL Number:MFCD00010914
 :112.41 g/mol
Merck Index:12,01649
Boiling Pt:765 °C (1013 hPa)
Melting Pt:321 °C
Density:8.642 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:2570
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09