Silver, powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-22-4
:
Formula:Ag
MDL Number:MFCD00003397
:107.87 g/mol
Boiling Pt:2212 °C (1013 hPa)
Melting Pt:961.9 °C
Density:10.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-131-3
Frequently Bought Together