Order Entry
United Kingdom
ContactUsLinkComponent
Gold, foil, Premion®, Thickness: 2,0 mm
Gold, foil, Premion®, Thickness: 2,0 mm
Catalog # 11387.FI
CAS Number:  
Gold, foil, Premion®, Thickness: 2,0 mm
Catalog # 11387.FI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    50×50 mm

Specifications

About this item

 :
 :196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436