Samarium, Sigma-Aldrich®
SIAL261211-10G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:150.36 g/mol
:12,08498
:1900 °C (1013 hPa)
:1076.85 °C
:7.42 g/cm³ (20 °C)
:Ambient
:231-128-7
:7440-19-9
:Sm
:MFCD00011233