Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-19-9
:
Formula:Sm
MDL Number:MFCD00011233
:150.36 g/mol
Merck Index:12,08498
Boiling Pt:1900 °C (1013 hPa)
Melting Pt:1076.85 °C
Density:7.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-128-7
Frequently Bought Together