
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Catalog # 35448.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Pk Info:25×25 mm
Specifications
About this item
:
:150.36 g/mol
Merck Index:12,08498
Boiling Pt:1900 °C (1013 hPa)
Melting Pt:1076.85 °C
Density:7.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-128-7
UN:3178
ADR:4.1,II
:GHS02
CAS Number:7440-19-9
Formula:Sm
MDL Number:MFCD00011233