
Samarium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-19-9
:
Formula:Sm
MDL Number:MFCD00011233
:150.36 g/mol
Merck Index:12,08498
ADR:4.1,II
:ampouled under argon
EINECS:231-128-7
UN:3089
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together