Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gadolinium
Gadolinium, Sigma-Aldrich®
Gadolinium, Sigma-Aldrich®
SIAL263060-5G
:
Merck
:
Gadolinium, Sigma-Aldrich®
SIAL263060-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
157.25 g/mol
:
M-Clarity™ quality level = MQ100
:
231-162-2
:
7440-54-2
:
Gd
:
MFCD00011022