Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-54-2
:
Formula:Gd
MDL Number:MFCD00011022
:157.25 g/mol
ADR:4.1,II
:Packaged under argon.
EINECS:231-162-2
UN:3178
:GHS02
Storage Temperature:Ambient
Frequently Bought Together