Erbium, Sigma-Aldrich®
Catalog # SIAL261084-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:167.26 g/mol
:M-Clarity™ quality level = MQ100
EINECS:231-160-1
CAS Number:7440-52-0
Formula:Er
MDL Number:MFCD00010987