Cerium, Sigma-Aldrich®
Catalog # SIAL461210-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:140.12 g/mol
Boiling Pt:3468 °C (1013 hPa)
Melting Pt:795 °C
Density:∼6.8 g/cm³ (20 °C)
EINECS:231-154-9
UN:1333
ADR:4.1,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
CAS Number:7440-45-1
Formula:Ce
MDL Number:MFCD00010924