Tellurium, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13494-80-9
:
Formula:Te
MDL Number:MFCD00134062
Boiling Pt:990
Melting Pt:450
Density:8.83 g/cm³ (20 °C)
Molecular Weight:127.6 g/mol
EINECS:236-813-4
UN:3284
ADR:6.1 III
Merck Index:12,09273
:M-Clarity™ quality level = MQ100
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together