Sulphur, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7704-34-9
:
Formula:S
MDL Number:MFCD00085316
Boiling Pt:445
Melting Pt:113…119 °C
Density:2.36 g/cm³ (20 °C)
Molecular Weight:32.07 g/mol
Flash Pt:168
Storage Temperature:Ambient
EINECS:231-722-6
UN:1350
ADR:4.1 III
Merck Index:13,09059
:M-Clarity™ quality level = MQ100
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together