Bismuth, powder, Sigma-Aldrich®
Catalog # SIAL264008-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:208.98 g/mol
Density:9.78 g/cm³ (20 °C)
UN:3089
ADR:4.1,III
:M-Clarity™ quality level = MQ100
EINECS:231-177-4
CAS Number:7440-69-9
Formula:Bi
MDL Number:MFCD00134033