Tin Reag. Ph. Eur. analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ300
:118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:232 °C
- Molar Mass:118.71 g/mol
- Boiling Point:2260 °C (1013 hPa)
- Density:7.3 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together