Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Lead, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIAL695912-25G
Health
Irritant
Environment
Supplier:   Merck
Lead, Sigma-Aldrich®
Catalog # SIAL695912-25G
Supplier:  Merck
CAS Number:  7439-92-1
Lead, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  7439-92-1
Price
£199.00
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications