Indium ≥99.9995% (metals basis), polycrystalline ingot, Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-74-6
:
Formula:In
MDL Number:MFCD00134048
:114.82 g/mol
Merck Index:13,04971
Boiling Pt:2072 °C
Melting Pt:156.6 °C
Density:7.310
Storage Temperature:Ambient
EINECS:231-180-0
Frequently Bought Together