Strontium, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-24-6
:
Formula:Sr
MDL Number:MFCD00134060
:87.62 g/mol
Merck Index:12,08994
Boiling Pt:1151 °C (1013 hPa)
Melting Pt:570…660 °C
Density:2.38 g/cm³ (770 °C)
UN:1393
ADR:4.3,II
:M-Clarity™ quality level = MQ100
EINECS:231-133-4
Frequently Bought Together