Strontium ≥99.8% (metals basis), pieces, distilled dendritic
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-24-6
:
Formula:Sr
MDL Number:MFCD00134060
:87.62 g/mol
Merck Index:12,08994
Boiling Pt:1151 °C (1013 hPa)
Melting Pt:570…660 °C
Density:2.38 g/cm³ (770 °C)
UN:3208
ADR:4.3,II
:ampouled under argon
EINECS:231-133-4
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together