Calcium, Sigma-Aldrich®
Catalog # SIAL327387-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:40.08 g/mol
Merck Index:13,01644
Boiling Pt:1484 °C (1013 hPa)
Melting Pt:850 °C
Density:1.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-179-5
UN:1401
ADR:4.3,II
:M-Clarity™ quality level = MQ100
:GHS02
CAS Number:7440-70-2
Formula:Ca
MDL Number:MFCD00085314