Order Entry
United Kingdom
ContactUsLinkComponent
Beryllium, beads ∼ 19 mm (0.75 in) Ø
Beryllium, beads ∼ 19 mm (0.75 in) Ø
Catalog # 45120.KF
CAS Number:  
Beryllium, beads ∼ 19 mm (0.75 in) Ø
Catalog # 45120.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :9.01 g/mol
Merck Index:12,01201
Boiling Pt:2970 °C (1013 hPa)
Melting Pt:1278 °C
Density:1.85 g/cm³ (20 °C)
EINECS:231-150-7
 :GHS06
 :GHS08
CAS Number:7440-41-7
Formula:Be
MDL Number:MFCD00134032