Sodium, Sigma-Aldrich®
SIAL282065-450G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:450 g
- Seq:3620
:
:22.99 g/mol
:13,08641
:881 °C (1013 hPa)
:97.8 °C
:0.9 g/cm³ (20 °C)
:Ambient
:231-132-9
:1428
:4.3,I
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:7440-23-5
:Na
:MFCD00085307