Order Entry
United Kingdom
ContactUsLinkComponent
Sodium
Sodium
Catalog # ROTH4469.1
Supplier:  Roth Carl
CAS Number:  
Sodium
Catalog # ROTH4469.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

About this item

 :
 :22.99 g/mol
Merck Index:13,08641
Boiling Pt:881 °C (1013 hPa)
Melting Pt:97.8 °C
Density:0.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-132-9
UN:1428
ADR:4.3,I
 :GHS02
 :GHS05
CAS Number:7440-23-5
Formula:Na
MDL Number:MFCD00085307