Order Entry
United Kingdom
ContactUsLinkComponent
Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
Supplier:  Greisinger
Certificates
undefined
Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
Supplier:  Greisinger
Specifications
  • Catalog No:
  • GRSG610040
  • GRSG610041
  • DO saturation:
  • 0,0 - 100,0 %
  • 0,0 - 100,0 %
  • DO barometric pressure range:
  • 300 - 5000 hPa (Sensor: 500 - 2000 hPa)
  • 300 - 5000 hPa (Sensor: 500 - 2000 hPa)
  • DO calibration:
  • Quick calibration on air at the push of a button or 2-point / 3-point (Air +0 % and 100 %)
  • Quick calibration on air at the push of a button or 2-point / 3-point (Air +0 % and 100 %)
  • Temperature range:
  • 0,0...50,0 °C
  • 0,0...50,0 °C
  • Display:
  • With backlight
  • With backlight
  • Data logging capacity:
  • Cyclical: 10000, Single: 1000
  • Cyclical: 10000, Single: 1000