Protein kinase C (20-28) (myristoylated)
Catalog # ENZOBMLP2050500
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:0,5 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Protein kinase C (20-28) (myristoylated)
- Purity:>98% (HPLC)
- Formulation:TFA salt. Lyophilised.
Specifications
About this item
PKC inhibitor
Pseudosubstrate sequence from PKCalpha and ß. Highly specific inhibitor, which is N-terminal myristoylated to allow membrane permeability. IC50=8 µM for TPA activation of MARCKS phosphorylation in fibroblast primary cultures, and 98% inhibition at 100 µM. Partially reverses multidrug resistance in human breast cancer cells.