Order Entry
United Kingdom
ContactUsLinkComponent
Human recombinant ABL1, HIS-tag
Human recombinant ABL1, HIS-tag
Catalog # ENZOBMLSE5620005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant ABL1, HIS-tag
Catalog # ENZOBMLSE5620005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    ABL1
  • Purity:
    >70% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 150 mM sodium chloride, 0,25 mM DTT, 0,1 mM EGTA, 0,1 mM EDTA, 0,1 mM PMSF and 25% glycerol.

Specifications

About this item

Produced in insect cells.

The ABL1 protooncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in cell differentiation, cell division, cell adhesion, and stress responses. The c-Abl protein is inhibited by its SH3 domain, and deletion of this domain turns ABL1 into an oncogene. A chromosomal translocation results in the head-to-tail fusion of the BCR and ABL1 genes. This translocation is present in many cases of chronic myelogeneous leukemia. The DNA-binding activity of Abl1 is regulated by CDC2-mediated phosphorylation.