Order Entry
United Kingdom
ContactUsLinkComponent
LRRKtide
LRRKtide
Catalog # ENZOBMLP2670001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
LRRKtide
Catalog # ENZOBMLP2670001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    LRRKtide
  • Purity:
    >95% (HPLC)
  • Formulation:
    Lyophilised

Specifications

About this item

LRRK substrate

This peptide, RLGRDKYKTLRQIRQ, is derived from ezrin/moesin/radixin and is a substrate for LRRK kinase.