Order Entry
United Kingdom
ContactUsLinkComponent
CREBtide
CREBtide
Catalog # ENZOBMLP1950001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
CREBtide
Catalog # ENZOBMLP1950001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    CREBtide
  • Purity:
    95%

Specifications

About this item

PKA substrate

This peptide, KRREILSRRPSYR, is an excellent substrate for many serine kinases, including PKA and PKC.