Order Entry
United Kingdom
ContactUsLinkComponent
Human recombinant CD30:Fc
Human recombinant CD30:Fc
Catalog # ENZOALX522027C050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Human recombinant CD30:Fc
Catalog # ENZOALX522027C050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
  • Species:
    Human
  • Seq:
    02
  • Environmentally Preferable:
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    CD30:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The cysteine-rich region of human CD30 (Ki-1) (aa 1-380) is fused to the Fc portion of human IgG1 .