Order Entry
United Kingdom
ContactUsLinkComponent
GM1 Pentasaccharide . Sodium Salt
GM1 Pentasaccharide . Sodium Salt
Catalog # ENZOALX305006M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
GM1 Pentasaccharide . Sodium Salt
Catalog # ENZOALX305006M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    GM1 Pentasaccharide sodium salt
  • Purity:
    >98%

Specifications

About this item

Semisynthetic (from GM1).