Human humanin
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Bax inhibitor
Humanin was originally identified as an anti-apoptotic peptide of 24 aa encoded in a cDNA that rescued neuronal cells from apoptosis induced by presenilin mutants associated with familial Alzheimer's disease and by amyloid-beta protein. Mutagenesis studies showed that the cysteine at position 8 of the humanin peptide is critical for its anti-apoptotic function. It has now been shown that Bax (Bcl-2 associated X protein), an apoptosis-inducing protein, interacts with humanin. Humanin prevents the translocation of Bax from the cytosol to mitochondria. Humanin peptides also block Bax association with isolated mitochondria, and suppress cytochrome c release in vitro.
Specifications
- Species:Human
- Protein/peptide name:Humanin
- Purity:-
Specifications
Frequently Bought Together