Order Entry
United Kingdom
ContactUsLinkComponent
Myeloperoxidase (human), ELISA kit
Myeloperoxidase (human), ELISA kit
Catalog # ENZOADI900115
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Myeloperoxidase (human), ELISA kit
Catalog # ENZOADI900115
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
    Human
  • Description:
    Myeloperoxidase (human), ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    0,019 ng/ml (range 0,195 - 12,5 ng/ml)

Specifications

About this item

For the quantitative determination of human Myeloperoxidase of culture supernatants, nasal lavage, Lithium-heparin plasma, sputum supernatant, and urine.

The Myeloperoxidase (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Tihs sensitive kit lets you quantitate as little as 19 pg/ml MPO in a wide variety of sample matrices that have been validated in-house. Convenient ready-to-use liquid color-coded reagents reduce error and save time in the lab.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 31 concentrate, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.