6-keto-PGF1α ELISA Kits
Catalog # ENZOADI900004
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:6-keto-PGF1α
- Description:6-keto-PGF1α ELISA kit
- Seq:02
- Sensitivity:1,40 pg/ml (range 3,2 - 50,000 pg/ml)
Specifications
About this item
Highly sensitive ELISA, detecting as low as 1.40pg/ml in just 3 hours.
For the quantitative determination of 6-keto-PGF1α in culture supernatants, serum, saliva, and urine from any species.
The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 1α EIA kit has been fully validated in-house in a wide variety of sample matrices.
Ordering information: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-272 (5x96 wells).