NGAL (Dog) ELISA kit
Catalog # ENZOBPDKIT043
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:NGAL
- Description:NGAL (Dog) ELISA kit
- Seq:02
- Storage conditions:4 °C (Long term)
- Sensitivity:0,56 pg/ml (Range 4 - 400 pg/ml)
Specifications
About this item
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in dog samples. Absorbance is read at 450 nm.
- Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
- Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
For the quantitative determination of NGAL in dog urine, plasma, serum, tissue extracts, or culture supernatants.
Delivery: Kit consists of microwell plate coated with anti-NGAL (dog) antibody (96 wells), 5X sample diluent, NGAL Dog calibrators 1-8; 0, 4, 10, 20, 40, 100, 200, 400 pg/ml, 25X wash solution concentrate, biotinylated Dog NGAL antibody, HRP-Streptavidin, TMB substrate, stop solution.