Order Entry
United Kingdom
ContactUsLinkComponent
 
HSP70B' ELISA kit
  ENZOADIEKS725A
New Product
 :  
HSP70B' ELISA kit
  ENZOADIEKS725A
New Product
 :  

 

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
  • Description:
    HSP70B' ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Storage conditions:
    4 °C (long term), –20 °C (For standards)
  • Sensitivity:
    62 pg/ml

 

 

The HSP70B' EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 4.5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.Only commercially available ELISA kit for ultra-sensitive HSP70B' detection in cell lysates, serum, and tissue.

  • Only source for HSP70B’ ELISA, due to patent protection
  • Precious sample compared to Western blot analysis
  • Fast and quantitative results

For the quantitative determination of HSP70B' in cell lysates, serum, and tissue of human origin.

 : Kit includes microtiter plate, 5X extraction reagent, standard, sample diluent, wash buffer concentrate, antibody, antibody diluent, HRP conjugate, conjugate diluent, TMB substrate, stop solution 2.