Leptin (Rat), ELISA Kits
Catalog # ENZOADI900015A
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Conjugate ELISA:HRP-Streptavidin
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Leptin (LEP)
- Description:Leptin (rat), ELISA kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sample type:Culture supernatants, plasma, and serum
- Detection method:Colorimetric 450 nm
- Detection range:100 - 6400 pg/ml
- Sensitivity:67,2 pg/ml
- Regulatory status:RUO
- Tests per kit:96 wells
Specifications
About this item
This Leptin (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. For the quantitative determination of rat leptin in culture supernatants, plasma, and serum.
- No edge effect: all incubations at room temperature eliminate risk of evaporation at 37 °C
- Broad dynamic range: easily quantify high and low levels of leptin
- Convenient method
- Pre-coated plates and ready-to-use reagents
Delivery: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.