Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-HSP70 Rabbit Polyclonal Antibody
 
Anti-HSP70 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ENZOADISPA811F
  • ENZOADISPA811D
  • Antigen symbol:
  • HSP70
  • HSP70
  • Antigen name:
  • Heat Shock Protein 70
  • Heat Shock Protein 70
  • Antigen synonyms:
  • HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
  • HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Form:
  • Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified

 

Frequently Bought Together