Laboratory freezers, spark-free, ISU and LSF
Catalog # 471-0837
Supplier: GLEN DIMPLEX
Specifications
- W×D×H ext.:595×640×1570 mm
- Capacity:232 l
- Description:Upright freezer, LSF232
- Seq:04
- Model:LSF232
- Cat. No.:471-0837
- Alarm:Audible and visual for temperature
- Defrost:Manual
- Door lock:Yes
- Door type:Solid, reversible door
- Shelves/drawers:2 compartments with drop-down fronts and 4 solid drawers
- Weight:60 kg
- Pk:1
- Temperature:−18...−25 °C
Specifications
About this item
Lec laboratory spark-free freezers offer safe, reliable storage of volatile liquids, reagents, poisons and samples. They have spark-free interiors to reduce the risk of internal explosions, CFC- and HCFC-free refrigeration systems ensuring minimal risk to the environment and locks to prevent theft or unauthorised access.
- Manual defrost
- Temperature alarm
- Reversible door
Temperature range: −18 to −20 or −25 °C