GenSaver™ and GenSaver™ 2.0 collection cards are treated with a proprietary chemistry intended to prevent environmentally induced degradation. These cards allow long term preservation of DNA from biological fluids stored at ambient temperature.
- Chemical treatment of these cards allows long-term ambient protection of DNA from dried matrix spots and improved qualitative and quantitative DNA recovery
- Cards are manufactured in accordance to the world’s first international standard for forensic consumables, minimising the risk of human DNA contamination
- DNA preserved on these cards can be used, even after 20 years of ambient storage, to generate high quality next generation sequencing data
- GenSaver™ - Suitable for collection of blood samples
- GenSaver™ Color - Suitable for the collection of colourless biosamples, such as saliva, buccal cells and urine
GenSaver™ and GenSaver™ Color cards are designed for the collection, transport and storage at ambient temperature of DNA from biological fluids. The fibre-based material of these cards is made of pure absorbent fibres, impregnated with a proprietary chemical formulation intended to prevent environmentally induced degradation for long-term ambient preservation of DNA.
GenSaver™ 2.0 and GenSaver™ Color 2.0 collection cards offer additional features, preventing the growth of microorganisms during the long term ambient storage of DNA.
- Catalog No:
- BINZ8.566.0003.B-G
- BINZ8.566.0003.B-K
- BINZ8.566.0003.B-N
- BINZ8.567.0003.B-G
- BINZ8.567.0003.B-K
- BINZ8.567.0003.B-N
- 730-1487
- 730-1486
- 730-1349
- 730-0684
- 730-0662
- Model:
- GenSaver™ 2.0
- GenSaver™ 2.0
- GenSaver™ 2.0
- GenSaver™ Color 2.0
- GenSaver™ Color 2.0
- GenSaver™ Color 2.0
- GenSaver™ Color 2.0
- GenSaver™ Color 2.0
- GenSaver™ Color 2.0
- GenSaver™ 2.0
- GenSaver™ 2.0
- Max. volume/sample area:
- 125 μl
- 125 μl
- 125 μl
- 70 μl
- 70 μl
- 70 μl
- 70 μl
- 70 μl
- 70 μl
- 125 μl
- 125 μl
- Sample type:
- Blood
- Blood
- Blood
- Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Saliva
Buccal cells
Urine
Bacterial suspension
Cell lysate - Blood
- Blood
- Storage conditions:
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years
- Max storage : 20 years