Hydrochloric acid 0.01 M, Titripur®, Supelco®
Catalog # 1.60238.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Storage:Store at +15 °C to +25 °C
- Molarity:0,01 M
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:36.46 g/mol
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324