Order Entry
United Kingdom
ContactUsLinkComponent
 
Carbon nanotube multi-walled ≥90% (carbon basis), powder, Length: 5 - 9 µm, Ø 110 - 170 nm, Sigma-Aldrich®
Catalog # SIAL659258-10G
Supplier:  Merck
CAS Number:  
Carbon nanotube multi-walled ≥90% (carbon basis), powder, Length: 5 - 9 µm, Ø 110 - 170 nm, Sigma-Aldrich®
Catalog # SIAL659258-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Assay:
    90%
  • Form:
    Powder
  • Composition :
    carbon >90%
  • Diameter:
    110 – 170 nm
  • Density (25 °C):
    1,7 g/ml
  • Color:
    Black
  • Iron (Fe):
    ≤0,1%
  • Length:
    5 – 9 μm
  • Stabilty (inert atmosphere) :
    3697 °C

Specification Test Results

About this item

 :
 :12.011 g/mol
Melting Pt:3652…3697 °C (lit.)
Density:1.7 g/ml (25 °C)
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:308068-56-6
Formula:C
MDL Number:MFCD00133992