Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sterilisation wrap
Sterilisation wrap
Sterilisation wrap
Supplier:  DISTRIBUTOR
Certificates
Sterilisation wrap
2 Options Available Below
Product Details & Documents
Sterilisation wrap is available in embossed crepe or plain papers.
Certifications: Manufactured under BS EN ISO.9001.2008 and BS EN ISO 13485:2003 and conforms to BS EN ISO11607-1:2006 part 1 and BS EN 868:2009 part 2.
Product Family Options
Product Information
  • Colour
    Dimensions
    Availability
    Price
  • DIUKSSWC1315
    Sterilisation wrap, crepe paper, 60 gsm
    Sterilisation wrap, crepe paper, 60 gsm
    White
    130×150 mm
    Log in for availability
    Log in for availability
    1 * 125 items
    1 * 125 items - £246.00
    £246.00
    1 * 125 items
       £246.00
  • DIUKSSWP1315
    Sterilisation wrap, plain paper, 60 gsm
    Sterilisation wrap, plain paper, 60 gsm
    White
    130×150 mm
    Log in for availability
    Log in for availability
    1 * 125 items
    1 * 125 items - £276.00
    £276.00
    1 * 125 items
       £276.00